Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRS8
Entrez Gene ID
844341
Gene Name
FAR1-related sequence 8
Gene Aliases
AT1G80010, F18B13.10, F18B13_10, FAR1-related sequence 8
Chromosome Location
Chr.1: 30097336 - 30100045 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106650.3 NP_178118.2 2 - 2 2283 86

View Full Product Details