Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UVH3
Entrez Gene ID
822427
Gene Name
5'-3' exonuclease family protein
Gene Aliases
AT3G28030, ULTRAVIOLET HYPERSENSITIVE 3, UV REPAIR DEFECTIVE 1, UVR1
Chromosome Location
Chr.3: 10424179 - 10431364 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338928.1 NP_001319661.1 12 - 13 3211 121
NM_001338929.1 NP_001325749.1 12 - 13 3211 121
NM_001338930.1 NP_001325748.1 11 - 12 3215 121
NM_001338931.1 NP_001325750.1 9 - 10 2665 121

View Full Product Details