Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UVH3
Entrez Gene ID
822427
Gene Name
5'-3' exonuclease family protein
Gene Aliases
AT3G28030, ULTRAVIOLET HYPERSENSITIVE 3, UV REPAIR DEFECTIVE 1, UVR1
Chromosome Location
Chr.3: 10424179 - 10431364 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338928.1 NP_001319661.1 6 - 7 958 69
NM_001338929.1 NP_001325749.1 6 - 7 958 69
NM_001338930.1 NP_001325748.1 5 - 6 962 69
NM_001338931.1 NP_001325750.1 3 - 4 412 69

View Full Product Details