Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHOT1
Entrez Gene ID
823721
Gene Name
phototropin 1
Gene Aliases
AT3G45780, F16L2.3, JK224, NONPHOTOTROPIC HYPOCOTYL 1, NPH1, PHOTOTROPIN, ROOT PHOTOTROPISM 1, RPT1, phototropin 1
Chromosome Location
Chr.3: 16816721 - 16824210 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035737.2 NP_001030814.1 12 - 13 2318 85
NM_114447.4 NP_190164.1 13 - 14 2316 85
GenBank mRNA AF030864.1 - 13 - 14 2171 85
AF360218.1 - 12 - 13 2125 85
AY040062.1 - 12 - 13 2108 85

View Full Product Details