Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG16
Entrez Gene ID
828904
Gene Name
SET domain protein 16
Gene Aliases
AT4G27910, ATX4, SET domain protein 16, T13J8.20, T13J8_20
Chromosome Location
Chr.4: 13893915 - 13900630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118929.5 NP_194520.3 2 - 3 1612 74

View Full Product Details