Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAP7
Entrez Gene ID
832446
Gene Name
amino acid permease 7
Gene Aliases
AT5G23810, MRO11.15, MRO11_15, amino acid permease 7
Chromosome Location
Chr.5: 8028238 - 8030888 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036857.1 NP_001031934.1 2 - 3 402 115
NM_001343827.1 NP_001330892.1 2 - 3 542 115
NM_001343828.1 NP_001330893.1 2 - 3 542 115
NM_001343829.1 NP_001330894.1 2 - 3 542 115
NM_122286.4 NP_197770.1 2 - 3 542 115
GenBank mRNA AK316999.1 - 2 - 3 344 115

View Full Product Details