Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SQE6
Entrez Gene ID
832482
Gene Name
squalene monooxygenase 6
Gene Aliases
AT5G24160, K12G2.4, K12G2_4, squalene monoxygenase 6
Chromosome Location
Chr.5: 8182917 - 8186704 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343845.1 NP_001330191.1 2 - 3 370 66
NM_001343846.1 NP_001330190.1 1 - 2 276 66
NM_122321.5 NP_197804.1 2 - 3 412 66
GenBank mRNA AJ005927.1 - 2 - 3 343 66

View Full Product Details