Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR5
Entrez Gene ID
832518
Gene Name
two-component response regulator-like protein
Gene Aliases
APRR5, AT5G24470, T31K7.5, T31K7_5, pseudo-response regulator 5
Chromosome Location
Chr.5: 8356204 - 8358546 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122355.4 NP_568446.2 1 - 2 230 65
GenBank mRNA AY062114.1 - 1 - 2 350 65
AY143934.1 - 1 - 2 230 65

View Full Product Details