Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP71A14
Entrez Gene ID
832566
Gene Name
cytochrome P450, family 71, subfamily A, polypeptide 14
Gene Aliases
''cytochrome P450, AT5G24960, F6A4.170, F6A4_170, cytochrome P450, family 71, polypeptide 14, polypeptide 14'', subfamily A
Chromosome Location
Chr.5: 8599735 - 8603321 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343905.1 NP_001331829.1 1 - 2 515 148
NM_122405.2 NP_197878.1 1 - 2 515 148

View Full Product Details