Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71A14
- Entrez Gene ID
-
832566
- Gene Name
- cytochrome P450, family 71, subfamily A, polypeptide 14
- Gene Aliases
-
''cytochrome P450, AT5G24960, F6A4.170, F6A4_170, cytochrome P450, family 71, polypeptide 14, polypeptide 14'', subfamily A
- Chromosome Location
-
Chr.5: 8599735 - 8603321 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122405.2 |
NP_197878.1 |
3 - 4 |
1290 |
115 |
View Full Product Details