Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRO2
Entrez Gene ID
838965
Gene Name
similar to RCD one 2
Gene Aliases
AT1G23550, F28C11.18, similar to RCD one 2
Chromosome Location
Chr.1: 8350728 - 8352791 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102204.3 NP_173769.1 1 - 2 673 125
GenBank mRNA BT022003.1 - 1 - 2 599 125

View Full Product Details