Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL3
Entrez Gene ID
839032
Gene Name
SIN3-like 3
Gene Aliases
ARABIDOPSIS THALIANA SIN3 HOMOLOG, AT1G24190, ATSIN3, F3I6.12, F3I6_12, SIN3, SIN3-like 3
Chromosome Location
Chr.1: 8563510 - 8570580 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198151.2 NP_001185080.1 17 - 18 3508 120
NM_001332641.1 NP_001323398.1 17 - 18 3464 120
NM_102265.3 NP_173829.3 17 - 18 3445 120

View Full Product Details