Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNL3
- Entrez Gene ID
-
839032
- Gene Name
- SIN3-like 3
- Gene Aliases
-
ARABIDOPSIS THALIANA SIN3 HOMOLOG, AT1G24190, ATSIN3, F3I6.12, F3I6_12, SIN3, SIN3-like 3
- Chromosome Location
-
Chr.1: 8563510 - 8570580 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102265.3 |
NP_173829.3 |
8 - 9 |
1216 |
133 |
View Full Product Details