Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP4
Entrez Gene ID
839730
Gene Name
receptor like protein 4
Gene Aliases
AT1G28340, AtRLP4, F3M18.23, F3M18_23, receptor like protein 4
Chromosome Location
Chr.1: 9939961 - 9943616 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102600.2 NP_174156.1 1 - 2 292 92

View Full Product Details