Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MKK9
Entrez Gene ID
843685
Gene Name
MAP kinase kinase 9
Gene Aliases
AT1G73500, ATMKK9, MAP kinase kinase 9, T9L24.32, T9L24_32
Chromosome Location
Chr.1: 27639091 - 27640643 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106009.3 NP_177492.1 1 - 1 430 58
GenBank mRNA AK118530.1 - 1 - 1 180 58
AY084571.1 - 1 - 1 316 58
BT005300.1 - 1 - 1 138 58

View Full Product Details