Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdc2cAt
Entrez Gene ID
833938
Gene Name
CDC2C
Gene Aliases
AT5G39420, CDC2C, MUL8.100, MUL8_100
Chromosome Location
Chr.5: 15771931 - 15775251 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123304.4 NP_198758.2 4 - 5 1456 76
GenBank mRNA BT046183.1 - 4 - 5 1155 76

View Full Product Details