Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSFA6A
Entrez Gene ID
834406
Gene Name
heat shock transcription factor A6A
Gene Aliases
AT-HSFA6A, AT5G43840, MQD19.20, MQD19_20, heat shock transcription factor A6A, heat shock transcription factor A6A
Chromosome Location
Chr.5: 17625353 - 17626487 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123751.2 NP_199197.1 1 - 2 360 110
GenBank mRNA DQ447035.1 - 1 - 2 237 110

View Full Product Details