Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckl7
Entrez Gene ID
834433
Gene Name
casein kinase I-like 7
Gene Aliases
AT5G44100, MLN1.2, MLN1_2, casein kinase I-like 7
Chromosome Location
Chr.5: 17749057 - 17752849 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344533.1 NP_001330873.1 13 - 14 1427 86
NM_123777.4 NP_199223.1 12 - 13 1551 86
GenBank mRNA AY943846.1 - 12 - 13 997 86
BT015795.1 - 12 - 13 997 86
BT021097.1 - 12 - 13 1093 86

View Full Product Details