Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXO70A3
- Entrez Gene ID
-
835311
- Gene Name
- exocyst subunit exo70 family protein A3
- Gene Aliases
-
AT5G52350, ATEXO70A3, K24M7.8, K24M7_8, exocyst subunit exo70 family protein A3
- Chromosome Location
-
Chr.5: 21254911 - 21257618 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124614.2 |
NP_200048.2 |
8 - 9 |
1636 |
95 |
View Full Product Details