Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB23
Entrez Gene ID
818166
Gene Name
plant U-box 23
Gene Aliases
AT2G35930, AtPUB23, F11F19.16, F11F19_16, plant U-box 23
Chromosome Location
Chr.2: 15082855 - 15084559 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129152.3 NP_181137.2 1 - 1 1044 99
GenBank mRNA AK227343.1 - 1 - 1 958 99
BT005829.1 - 1 - 1 821 99

Gene Symbol
AT2G35935
Entrez Gene ID
28718330
Gene Name
hypothetical protein
Gene Aliases
AT2G35935
Chromosome Location
Chr.2: 15083353 - 15084340 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336583.1 NP_001324636.1 1 - 1 164 99

View Full Product Details