Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OFP15
- Entrez Gene ID
-
818178
- Gene Name
- ovate family protein 15
- Gene Aliases
-
ARABIDOPSIS THALIANA OVATE FAMILY PROTEIN 15, AT2G36050, ATOFP15, F11F19.32, F11F19_32, ovate family protein 15
- Chromosome Location
-
Chr.2: 15135568 - 15137012 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129164.3 |
NP_565833.1 |
1 - 1 |
1324 |
177 |
View Full Product Details