Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PECT1
Entrez Gene ID
818449
Gene Name
phosphorylethanolamine cytidylyltransferase 1
Gene Aliases
AT2G38670, T6A23.13, T6A23_13, phosphorylethanolamine cytidylyltransferase 1
Chromosome Location
Chr.2: 16168712 - 16171918 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129424.5 NP_181401.1 8 - 9 1451 76
GenBank mRNA AY092984.1 - 8 - 9 1291 76
AY128807.1 - 8 - 9 1184 76

View Full Product Details