Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAO2
Entrez Gene ID
818904
Gene Name
polyamine oxidase 2
Gene Aliases
AT2G43020, ATPAO2, MFL8.12, MFL8_12, polyamine oxidase 2
Chromosome Location
Chr.2: 17891503 - 17894635 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129863.3 NP_181830.1 6 - 7 975 100
GenBank mRNA AF364952.1 - 6 - 7 893 100
BT029025.1 - 6 - 7 533 100

View Full Product Details