Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDH2-3
Entrez Gene ID
836640
Gene Name
succinate dehydrogenase 2-3
Gene Aliases
AT5G65165, succinate dehydrogenase 2-3
Chromosome Location
Chr.5: 26034422 - 26036057 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345673.1 NP_001331888.1 1 - 2 311 123
NM_148160.3 NP_680465.2 1 - 2 311 123
GenBank mRNA AJ278912.1 - 1 - 2 181 123

View Full Product Details