Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADF3
Entrez Gene ID
836110
Gene Name
actin depolymerizing factor 3
Gene Aliases
ACTIN DEPOLYMERIZING FACTOR 3, AT5G59880, MMN10.4, MMN10_4, actin depolymerizing factor 3
Chromosome Location
Chr.5: 24120164 - 24121949 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180896.3 NP_851227.1 2 - 3 487 96
GenBank mRNA AF102821.1 - 2 - 3 356 96
AF360169.1 - 2 - 3 358 96
AY050334.1 - 2 - 3 420 96
AY056345.1 - 1 - 2 269 96
AY086069.1 - 2 - 3 380 96
AY094033.1 - 1 - 2 269 96

View Full Product Details