Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT3G14040
Entrez Gene ID
820618
Gene Name
Pectin lyase-like superfamily protein
Gene Aliases
AT3G14040
Chromosome Location
Chr.3: 4648236 - 4650319 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112258.4 NP_188020.1 1 - 2 358 162

View Full Product Details