Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHC1A
Entrez Gene ID
818680
Gene Name
RING-H2 finger C1A
Gene Aliases
AT2G40830, RING-H2 FINGER PROTEIN RHC1A, RING-H2 finger C1A, T20B5.3, T20B5_3
Chromosome Location
Chr.2: 17042623 - 17044915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336872.1 NP_001324188.1 2 - 2 1808 69
NM_129646.5 NP_565942.1 2 - 2 1165 69
NM_201922.3 NP_973651.1 2 - 2 1274 69
NM_201923.4 NP_973652.1 3 - 3 1177 69
GenBank mRNA AF079180.1 - 2 - 2 1073 69
AY139987.1 - 2 - 2 1084 69
BT008726.1 - 1 - 1 886 69

View Full Product Details