Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPB6A
Entrez Gene ID
835269
Gene Name
RNA polymerase Rpb6
Gene Aliases
AT5G51940, MSG15.2, MSG15_2, NRPD6A, NRPE6A
Chromosome Location
Chr.5: 21104551 - 21105947 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124573.3 NP_200007.1 2 - 3 201 147
GenBank mRNA AF370301.1 - 2 - 3 160 147
AY063088.1 - 2 - 3 73 147
AY088602.1 - 2 - 3 166 147

View Full Product Details