Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBR1
Entrez Gene ID
505142
Gene Name
T-box brain transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 34960314 - 34968013 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192049.1 NP_001178978.1 1 - 2 783 86

View Full Product Details