Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLR6
Entrez Gene ID
407237
Gene Name
toll like receptor 6
Gene Aliases
TLR1
Chromosome Location
Chr.6: 58050661 - 58091639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001159.1 NP_001001159.1 2 - 3 151 131
GenBank mRNA AJ618974.1 - 2 - 3 151 131
AM086205.1 - 2 - 3 119 131

View Full Product Details