Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAT5A
Entrez Gene ID
282375
Gene Name
signal transducer and activator of transcription 5A
Gene Aliases
STAT5A/MGF, STATA5
Chromosome Location
Chr.19: 42395537 - 42415495 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012673.1 NP_001012691.1 1 - 2 63 66
GenBank mRNA Z72482.1 - 1 - 2 63 66

View Full Product Details