Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYGB
Entrez Gene ID
281720
Gene Name
crystallin gamma B
Gene Aliases
-
Chromosome Location
Chr.2: 96417868 - 96420083 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013594.1 NP_001013612.1 1 - 2 34 56
GenBank mRNA EF208024.1 - 1 - 2 27 56
M16894.1 - 1 - 2 34 56
X01036.1 - - 30 56

View Full Product Details