Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM76A
Entrez Gene ID
535402
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
Chromosome Location
Chr.2: 125604199 - 125635148 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015660.1 NP_001015660.1 8 - 9 879 106
GenBank mRNA BT020680.1 - 8 - 9 879 106

View Full Product Details