Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CISD1
Entrez Gene ID
531444
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
-
Chromosome Location
Chr.26: 745914 - 764106 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034057.1 NP_001029229.1 2 - 3 347 85
GenBank mRNA BC103105.1 - 2 - 3 347 85

View Full Product Details