Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTN
Entrez Gene ID
507525
Gene Name
vitronectin
Gene Aliases
-
Chromosome Location
Chr.19: 19855911 - 19858842 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035050.2 NP_001030222.1 7 - 8 1416 94
GenBank mRNA BC103132.1 - 7 - 8 1418 94

View Full Product Details