Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDX4
Entrez Gene ID
281999
Gene Name
peroxiredoxin 4
Gene Aliases
-
Chromosome Location
Chr.X: 119515820 - 119528785 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174433.2 NP_776858.1 2 - 3 459 95
GenBank mRNA AF305563.1 - 2 - 3 459 95
BC109824.1 - 2 - 3 426 95

View Full Product Details