Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCKDHA
Entrez Gene ID
282149
Gene Name
branched chain keto acid dehydrogenase E1 subunit alpha
Gene Aliases
-
Chromosome Location
Chr.18: 50541524 - 50560679 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174506.1 NP_776931.1 6 - 7 982 75
GenBank mRNA BC109669.1 - 6 - 7 876 75
J03759.1 - 6 - 7 982 75

View Full Product Details