Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD40LG
Entrez Gene ID
282387
Gene Name
CD40 ligand
Gene Aliases
TNFSF5
Chromosome Location
Chr.Unknown: 8251624 - 8263447 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174624.2 1 - 2 171 127
GenBank mRNA Z48469.1 - - 171 127

View Full Product Details