Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BYSL
Entrez Gene ID
514128
Gene Name
bystin like
Gene Aliases
-
Chromosome Location
Chr.23: 15688030 - 15697325 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014924.1 NP_001014924.1 2 - 3 699 92
GenBank mRNA BC151370.1 - 2 - 3 752 92
BT020890.1 - 2 - 3 699 92

View Full Product Details