Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
540083
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
-
Chromosome Location
Chr.8: 59277545 - 59286524 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014959.1 NP_001014959.1 1 - 2 103 122
GenBank mRNA BT021176.1 - 1 - 2 103 122

View Full Product Details