Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAX1
Entrez Gene ID
540424
Gene Name
lymphocyte transmembrane adaptor 1
Gene Aliases
LAX
Chromosome Location
Chr.16: 1509969 - 1518575 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015674.1 NP_001015674.1 5 - 6 711 105
GenBank mRNA BT021859.1 - 5 - 6 711 105

View Full Product Details