Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORO1B
Entrez Gene ID
510128
Gene Name
coronin 1B
Gene Aliases
-
Chromosome Location
Chr.29: 45329804 - 45335272 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113243.1 NP_001106714.1 10 - 11 1418 76
GenBank mRNA BC103376.1 - 10 - 11 1400 76
BT021806.1 - 10 - 11 1389 76

View Full Product Details