Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FHL3
Entrez Gene ID
504795
Gene Name
four and a half LIM domains 3
Gene Aliases
-
Chromosome Location
Chr.3: 107958890 - 107967123 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034223.2 NP_001029395.1 4 - 5 691 85
GenBank mRNA BC103276.1 - 4 - 5 692 85

View Full Product Details