The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035495.1 | NP_001030572.1 | 1 - 2 | 324 | 80 |
GenBank mRNA | BT021820.1 | - | 1 - 2 | 324 | 80 |