Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK2B
Entrez Gene ID
617391
Gene Name
divergent protein kinase domain 2B
Gene Aliases
CXHXorf36, CXORF36
Chromosome Location
Chr.X: 98556745 - 98608340 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035495.1 NP_001030572.1 4 - 5 1057 131
GenBank mRNA BT021820.1 - 4 - 5 1057 131

View Full Product Details