Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCA1A
Entrez Gene ID
282243
Gene Name
guanylate cyclase activator 1A
Gene Aliases
-
Chromosome Location
Chr.23: 15891989 - 15898940 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174546.2 NP_776971.1 2 - 3 559 124
GenBank mRNA X95352.1 - - 586 124

View Full Product Details