Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK2
Entrez Gene ID
282590
Gene Name
potassium two pore domain channel subfamily K member 2
Gene Aliases
-
Chromosome Location
Chr.16: 68369619 - 68595681 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174686.2 NP_777111.1 7 - 8 1460 114
GenBank mRNA AY148474.1 - 7 - 8 1460 114

View Full Product Details