Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB27B
Entrez Gene ID
282858
Gene Name
RAB27B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.24: 54193395 - 54254695 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174738.2 NP_777163.1 3 - 4 557 73
GenBank mRNA AY010899.1 - 3 - 4 557 73
BC122732.1 - 4 - 5 403 73

View Full Product Details