Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4A1
Entrez Gene ID
504958
Gene Name
eukaryotic translation initiation factor 4A1
Gene Aliases
-
Chromosome Location
Chr.19: 27305544 - 27311552 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034228.1 NP_001029400.1 10 - 11 1095 76
GenBank mRNA BC103130.1 - 10 - 11 1095 76

View Full Product Details