Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4A1
Entrez Gene ID
504958
Gene Name
eukaryotic translation initiation factor 4A1
Gene Aliases
-
Chromosome Location
Chr.19: 27305544 - 27311552 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034228.1 NP_001029400.1 1 - 2 36 78
GenBank mRNA BC103130.1 - 1 - 2 36 78

View Full Product Details